missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELF4 Monoclonal Antibody (OTI4A3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527251
This item is not returnable.
View return policy
Description
ELF4 Monoclonal Antibody for Western Blot, IHC (P)
ELF4 is transcriptional activator that binds to DNA sequences containing the consensus 5'-WGGA-3'. The protein acts synergistically with RUNX1 to transactivate the IL3 promoter. Also transactivates the PRF1 promoter in natural killer (NK) cells. This protein plays a role in the development and function of NK and NK T-cells and in innate immunity.
Specifications
| ELF4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99607 | |
| Elf4 | |
| Human recombinant protein fragment corresponding to amino acids 83-345 of ELF4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4A3 | |
| Unconjugated | |
| Elf4 | |
| AV314029; BC042423; E74 like ETS transcription factor 4; E74-like factor 4; E74-like factor 4 (ets domain transcription factor); E74-like factor 4; myeloid elf-1-like factor; myeloid elf-1 like factor; ELF4; ELFR; ENSMUSG00000053512; ETS-related transcription factor Elf-4; Gm9907; MEF; myeloid elf-1 like factor; myeloid Elf-1-like factor; RGD1560743 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2000 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction