missing translation for 'onlineSavingsMsg'
Learn More

Elf4/MEF Antibody, Novus Biologicals™

Product Code. 18324887 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18324887 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18324887 Supplier Novus Biologicals Supplier No. H00002000D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Elf4/MEF Polyclonal antibody specifically detects Elf4/MEF in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Elf4/MEF
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.4)
Gene Accession No. NP_001412.1
Gene Alias E74-like factor 4, E74-like factor 4 (ets domain transcription factor), ETS-related transcription factor Elf-4, MEFELFRMyeloid Elf-1-like factor
Host Species Rabbit
Immunogen ELF4 (NP_001412.1, 1 a.a. - 663 a.a.) full-length human protein. MAITLQPSDLIFEFASNGMDDDIHQLEDPSVFPAVIVEQVPYPDLLHLYSGLELDDVHNGIITDGTLCMTQDQILEGSFLLTDDNEATSHTMSTAEVLLNMESPSDILDEKQIFSTSEMLPDSDPAPAVTLPNYLFPASEPDALNRAGDTSDQEGHSLEEKASREESAKKTGKSKKRIRKTKGNRSTSPVTDPSIPIRKKSKDGKGSTIYLWEFLLALLQDRNTCPKYIKWTQREKGIFKLVDSKAVSKLWGKQKNKPDMNYETMGRALRYYYQRGILAKVEGQRLVYQFKEMPKDLVVIEDEDESSEATAAPPQASTASVASASTTRRTSSRVSSRSAPQGKGSSSWEKPKIQHVGLQPSASLELGPSLDEEIPTTSTMLVSPAEGQVKLTKAVSASSVPSNIHLGVAPVGSGSALTLQTIPLTTVLTNGPPASTTAPTQLVLQSVPAASTFKDTFTLQASFPLNASFQDSQVAAPGAPLILSGLPQLLAGANRPTNPAPPTVTGAGPAGPSSQPPGTVIAAFIRTSGTTAAPRVKEGPLRSSSYVQGMVTGAPMEGLLVPEETLRELLRDQAHLQPLPTQVVSRGSHNPSLLGNQTLSPPSRPTVGLTPVAELELSSGSGSLLMAEPSVTTSGSLLTRSPTPAPFSPFNPTSLIKMEPHDI
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2000
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.