missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELF3 Recombinant Rabbit Monoclonal Antibody (7O2V4)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA535683

Product Code. 17126343

  • £437.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MAATCEISNI FSNYFSAMYS SEDSTLASVP PAATFGADDL VLTLSNPQMS LEGTEKASWL GEQPQFWSKT QVLDWISYQV EKNKYDASAI DFSRCDMDGA TLCNCALEEL RLVFGPLGDQ LHAQLRDLTS SSSDELSWII ELLEKDGMAF QEALDPGPFD QGSPFAQELL DDGQQASPYH PGSCGAGAPS PGSSDVSTAG TGASRSSHSS DSGGSDVDLD PTDGKLFPSD GFRDCKKGDP KHGKRKRGRP RKLSKEYWDC LEGKKSKHAP RGTHLWEFIR DILIHPELNE GLMKWENRHE GVFKFLRSEA VAQLWGQKKK NSNMTYEKLS RAMRYYYKRE ILERVDGRRL VYKFGKNSSG WKEEEVLQSR N.

ESE-1 (Epithelium-Specific Ets) is a novel, highly tissue-restricted member of the ets transcription factor/oncogene family, which has features distinct from those of any other ets-related factor. ESE-1 binds with high affinity to and transactivates the ets binding site in the promoter of the keratinocyte terminal differentiation marker gene, SPRR2A. ESE-1 is a critical regulator of epithelial cell differentiation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ELF3
Recombinant Monoclonal
0.8 mg/mL
PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
P78545, Q3UPW2, Q4V7E1
ELF3
Recombinant protein (or fragment).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ChIP Assay, ELISA, Western Blot, CUT&RUN
7O2V4
Unconjugated
ELF3
E74 like ETS transcription factor 3; E74-like factor 3; E74-like factor 3 (ets domain transcription factor); E74-like factor 3 (ets domain transcription factor, epithelial-specific ); E74-like factor 3 (ETS domain transcription factor, serine box, epithelial-specific); ELF3; elf3 {ECO:0000312; EMBL:AAH97980.1}; Epithelial-restricted with serine box; epithelium-restricted Ets protein ESX; Epithelium-specific Ets transcription factor 1; EPR-1; Ert; ESE-1; ESX; ets domain transcription factor, serine box (epithelial-specific); ETS-related transcription factor Elf-3; JEN; RP11-510N19.1
Rabbit
Affinity Chromatography
RUO
13710, 1999, 304815
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.