missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELF3/ESE-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-87945
This item is not returnable.
View return policy
Description
ELF3/ESE-1 Polyclonal specifically detects ELF3/ESE-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ELF3/ESE-1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
| P78545 | |
| ELF3 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ATCEISNIFSNYFSAMYSSEDSTLASVPPAATFGADDLVLTLSNPQMSLEGTEKASWLGEQPQFWSKTQVLDWISYQVEKNKYDASAIDFSRCDMDGATLCNCALEELRLVFGPLGDQLHAQLRDLTSSSSDELSWIIELLEKDGMA | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| E74-like factor 3, E74-like factor 3 (ets domain transcription factor, epithelial-specific ), Epithelial-restricted with serine box, epithelial-specific), Epithelium-restricted Ets protein ESX, Epithelium-specific Ets transcription factor 1, EPR-1, ets domain transcription factor, serine box (epithelial-specific), ETS-related transcription factor Elf-3, JEN, serine box | |
| Rabbit | |
| 41 kDa | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Chromatin Research, Transcription Factors and Regulators | |
| 1999 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction