missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596697
This item is not returnable.
View return policy
Description
Immunogen sequence: MATSLHEGPT NQLDLLIRAV EASVHSSNAH CTDKTIEAAE ALLHMESPTC LRDSRSPVEV FVPPCVSTPE FIHAAMRPDV ITETVVEVST EESEPMDTSP IPTSPDSHEP MKKKKVGRKP KTQQSPISNG SPELGIKKKP; Positive Samples: HL-60, A-549, MCF7, Jurkat; Cellular Location: Nucleus.
The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kDa. This gene encodes the 36 kD subunit. This subunit can interact with the C-terminal region of PCNA. It forms a core complex with the 38 and 40 kDa subunits. The core complex possesses DNA-dependent ATPase activity, which was found to be stimulated by PCNA in an in vitro system. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
| ELF2 | |
| Polyclonal | |
| Unconjugated | |
| ELF2 | |
| 2610036A20Rik; A230104O07Rik; AW111824; BB183398; E74 like ETS transcription factor 2; E74-like factor 2; E74-like factor 2 (ets domain transcription factor); E74-like factor 2 (ets domain transcription factor) isoform 1; E74-like factor 2 (ets domain transcription factor) isoform 2; elf2; ets domain transcription factor; ets family transcription factor ELF2A2; ets family transcription factor ELF2C; ETS-related transcription factor Elf-2; EU32; NERF; NERF-1A; NERF-1a,b; NERF-1B; NERF-2; new Ets-related factor | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1998, 69257 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15723, Q9JHC9 | |
| ELF2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human ELF2 (NP_0068651). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction