Learn More
Invitrogen™ ELF1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5115174
Product Code. 17297933
  • £454.00 / 100µL
Description
Antibody detects endogenous levels of total ELF1.
This gene encodes an E26 transformation-specific related transcription factor. The encoded protein is primarily expressed in lymphoid cells and acts as both an enhancer and a repressor to regulate transcription of various genes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ELF1
Polyclonal
Unconjugated
ELF1
E74 like ETS transcription factor 1; E74-like factor 1; E74-like factor 1 (ets domain transcription factor); ELF 1; ELF1; Elf-1; ETS-like transcription factor Elf-1-like protein; ETS-related transcription factor Elf-1; ETS-related transcription factor elf-1-like protein; mElf 1; mElf-1; OTTHUMP00000018310; p70; Sts1
Rabbit
Affinity chromatography
RUO
1997
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide
P32519
ELF1
A synthesized peptide derived from human ELF1(Accession P32519), corresponding to amino acid residues I553-T603.