Learn More
ELF1 Mouse anti-Human, Unconjugated, Clone: 2D4A11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67138-1-IG-150UL
This item is not returnable.
View return policy
Description
The ELF1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ELF1. This antibody recognizes human antigen. The ELF1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ELF1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 2D4A11 | |
| Unconjugated | |
| 150 μL | |
| 1997 | |
| IgG1 |