missing translation for 'onlineSavingsMsg'
Learn More

Eleutheroside E, MedChemExpress

Product Code. 16489036
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489036 100 mg 100mg
16468928 10 mg 10mg
16451368 5 mg 5mg
16414807 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16489036 Supplier Medchem Express Supplier No. HYN0272
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Eleutheroside E, a principal component of Eleutherococcus enticosus, has anti-inflammatory and protective effects in ischemia heart.

Eleutheroside E, a principal component of Eleutherococcus enticosus, has anti-inflammatory and protective effects in ischemia heart.

Specifications

Chemical Name or Material Eleutheroside E
CAS 39432-56-9
Color White
Purity Grade Notes Research
Molecular Formula C34H46O18
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (134.64 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.35 mM; Need ultrasonic)
SMILES COC(C=C1[C@H]2[C@@](CO[C@@H]3C4=CC(OC)=C(O[C@@H]([C@@H]([C@@H](O)[C@@H]5O)O)O[C@@H]5CO)C(OC)=C4)([H])[C@]3([H])CO2)=C(C(OC)=C1)O[C@@H]([C@@H]([C@@H](O)[C@@H]6O)O)O[C@@H]6CO
Molecular Weight (g/mol) 742.72
Formula Weight 742.72
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.