missing translation for 'onlineSavingsMsg'
Learn More

Eletriptan N-Oxide, TRC

Product Code. 30180337
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180337 5 mg 5mg
30180312 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180337 Supplier Toronto Research Chemicals Supplier No. TRCE5050105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1217641-89-8
Molecular Formula C22H26N2O3S
Molecular Weight (g/mol) 398.52
Synonym 3-[[(2R)-1-Methyl-2-pyrrolidinyl]methyl]-5-[2-(phenylsulfonyl)ethyl]-1H-indole N Oxide;
IUPAC Name 5-[2-(benzenesulfonyl)ethyl]-3-[[(2R)-1-methyl-1-oxidopyrrolidin-1-ium-2-yl]methyl]-1H-indole
SMILES C[N+]1([O-])CCC[C@@H]1Cc2c[nH]c3ccc(CCS(=O)(=O)c4ccccc4)cc23

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C22H26N2O3S/c1-24(25)12-5-6-19(24)15-18-16-23-22-10-9-17(14-21(18)22)11-13-28(26,27)20-7-3-2-4-8-20/h2-4,7-10,14,16,19,23H,5-6,11-13,15H2,1H3/t19-,24?/m1/s1
Formula Weight 398.1664
Chemical Name or Material Eletriptan N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.