missing translation for 'onlineSavingsMsg'
Learn More

Elemicin, TRC

Product Code. 30012204
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012204 250 mg 250mg
30006270 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012204 Supplier Toronto Research Chemicals Supplier No. TRCE501020250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 487-11-6
Molecular Formula C12 H16 O3
Molecular Weight (g/mol) 208.25
Synonym Benzene, 1,2,3-trimethoxy-5-(2-propen-1-yl)-, Benzene, 1,2,3-trimethoxy-5-(2-propenyl)- (9CI), Benzene, 5-allyl-1,2,3-trimethoxy- (8CI), Elemicin (6CI), 1,2,3-Trimethoxy-5-(2-propen-1-yl)benzene, 1,2,3-Trimethoxy-5-(2-propenyl)benzene, 1-(3,4,5-Trimethoxyphenyl)-2-propene, 1-Allyl-3,4,5-trimethoxybenzene, 3,4,5-Trimethoxyallylbenzene, 4-Allyl-1,2,6-trimethoxybenzene, 5-(Prop-2-enyl)-1,2,3-trimethoxybenzene, 5-Allyl-1,2,3-trimethoxybenzene, NSC 16704
IUPAC Name 1,2,3-trimethoxy-5-prop-2-enylbenzene
SMILES COc1cc(CC=C)cc(OC)c1OC

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H16O3/c1-5-6-9-7-10(13-2)12(15-4)11(8-9)14-3/h5,7-8H,1,6H2,2-4H3
Formula Weight 208.1099
Chemical Name or Material Elemicin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.