Learn More
Honeywell™ ELECTROSOFT COMPOSITE Class 0 Latex Gloves
Product Code. 12697603
Change view
Click to view available options
Size:
8
9
10
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12697603 9 Each
12687603 8 Each
12657603 10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12697603 Supplier Honeywell™ Supplier No. 209200209
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Honeywell™ ELECTROSOFT COMPOSITE Class 0 Latex Gloves for electrical work are designed to be used in Arc Flash environments.
  • Insulating glove for electrical work
  • External layer in orange polychloroprene
  • Chlorinated inside
  • Category RC
  • ATPV 21.6/cal
  • Dielectric natural latex composition on chlorinated interior base makes it very comfortable
  • Product appreciated for its long life and precision
  • Patterned finish gives it a highly appreciated grip in damp conditions
  • The outer orange elastomer layer provides excellent mechanical resistance
  • Its anatomical shape makes flexing easier while ensuring a high level of protection
  • Specifically designed to be used in Arc Flash environement
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 60903-2003, CEI 903-2002, IEC 61482-1-2
Coating Material Latex, Polychloroprene
Color Orange
Size 9
Thickness (Metric) 1.1 mm
Length (Metric) 36 cm