missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ ELECTROSOFT Class 0 Latex Gloves
Honeywell™ ELECTROSOFT Class 0 Latex Gloves are ideal for electrical work and should be used with leather protectors for mechanical protection.
Brand: Honeywell™ 2091907-11
Code : NEW
Additional Details : Weight : 0.23400kg
Description
- Beige natural latex insulating glove
- Cut edge
- Category RC
- Thickness 1 mm
- Ergonomic shape and lightly powdered interior make the glove easy to slip on and off
- Natural latex base offers high dielectric characteristics
- Thickness ensures good dexterity while offering protection up to 1000 volts
- To be used with leather protectors for mechanical protection
Specifications
1 mm | |
11 | |
36 cm |
Latex | |
Beige | |
EN 60903 : 2003, CEI 903 : 2002 |