Learn More
3M™ Electronic Test System Unit
Product Code. 17791669
Click to view available options
:
Electronic Test System Unit; For Use With (Application): Sterilization Monitoring; Model: 4308
Product Code. 17791669
Brand: 3M™ undefined
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Electronic Test System Unit is an integrated solution revolutionizing sterilization monitoring.
  • Service parts available to authorized entities only
  • Fully automatic with Pass/Fail Bowie-Dick type test result
  • Data download to PC software for capture and trending
Specifications
For Use With (Application) Sterilization Monitoring
Model 4308