missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Electrolyte For Barium ISE

Product Code. 11967721
Change view
Click to view available options
Quantity:
20 mL
Unit Size:
20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11967721 20 mL 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11967721 Supplier Mettler Toledo™ Supplier No. 51107892
  • ONLINE EXCLUSIVE PRICE
    £54.00
    £53.00 / 20mL
    Save £1.00
    1% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Description Electrolyte for Barium ISE, 20mL
Filling Solution Type Electrolyte
Packaging 20 mL
Quantity 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.