Learn More
Bel-Art™ Electrode Holder
Product Code. 15631244
Change view
Click to view available options
Unit Size:
Each
Product Code. unitSize
15631244 Each
1 options
Product Code. 15631244 Supplier Bel-Art™ Supplier No. F183171000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For multi-purpose flexible arm stand.
Snaps onto the end of the Flexible Gooseneck Arm affording three stepped wells for holding an electrode, a reference electrode and an ATC probe.
Specifications
Type Electrode Holder
For Use With (Application) Holding an electrode, a reference electrode and an ATC probe