Learn More
Pepsin-HCl Electrode Cleaning Solution, Reagecon™
Ideal for removing protein contamination from the membrane and diaphragm of pH electrodes. Pepsin-HCl Electrode Cleaning Solution, Reagecon™ is a solution of pepsin A in hydrochloric acid. Electrode elements should be soaked for approximately one hour to maintain optimal performance.
Brand: Reagecon ECS
Code : 04
Additional Details : Weight : 0.58000kg
Description
Pepsin A is an enzyme that breaks down proteins into smaller peptides. It is produced naturally in the human digestive system and is produced commercially for use in laboratories and in the food industry. Pepsin in hydrochloric acid is the preferred cleaning solution for removing protein contamination.Specifications
100°C | |
Pepsin A in Hydrochloric Acid | |
0°C | |
500 mL |
Light Orange | |
Electrode cleaning for protein contamination | |
Characteristic | |
Miscible in water |
Safety and Handling
- ELECTRODE CLEANING SOLUTION
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Pepsin A