Learn More
Faster™ Electrical Modular Stands for SafeFAST and FlowFAST Safety Cabinets
Click to view available options
For Use With (Equipment):
SafeFAST 109/209
SafeFAST 112/312
SafeFAST 115/315
SafeFAST 118/318
Includes:
Stand
Stand with Castors
Description
- Stand height can be changed at the push of a button to raise or lower the cabinet
- Stand height range is 780 to 1080 mm
- For use with SafeFAST Classic, Elite, Premium, and FlowFAST V Safety Cabinets
- Please note: If cabinent is moved, it is strongly recommended to recommisison due to possible airflow changes
Specifications
Specifications
| For Use With (Equipment) | SafeFAST 112/312 |
| Includes | Stand |
| For Use With (Application) | Electronic Cabinet Height Adjustment |