Learn More
Specac™ Electrical Heating Jacket
Click to view available options
Type:
Electrical Heating Jacket
Description
This Electrically Heated Jacket is designed for liquid and solid sampling in transmission from ambient to 250°C. It is ideal for static or flow sampling capabilities and is compatible with all FTIR instruments. It consists of a central heatable chamber with a front cover plate. Liquid and solid sample cells are placed within the central chamber.
Specifications
Specifications
| Type | Electrical Heating Jacket |
| Includes | A central heatable chamber with a front cover plate |