missing translation for 'onlineSavingsMsg'
Learn More

Eldecalcitol, TRC

Product Code. 30010248
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010248 1 mg 1mg
30010202 10 mg 10mg
30008002 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010248 Supplier Toronto Research Chemicals Supplier No. TRCE5555401MG
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104121-92-8
Molecular Formula C30 H50 O5
Molecular Weight (g/mol) 490.72
Synonym (1α,2β,3β,5Z,7E)-2-(3-Hydroxypropoxy)-9,10-secocholesta-5,7,10(19)-triene-1,3,25-triol, 2β-(3-Hydroxypropoxy)-1α,25-dihydroxyvitamin D3, ED 71, Edirol, (1R,2R,3R,5Z)-2-(3-Hydroxypropoxy)-4-methylene-5-[(2E)-2-[(1R,3aS,7aR)-octahydro-1-[(1R)-5-hydroxy-1,5-dimethylhexyl]-7a-methyl-4H-inden-4-ylidene]ethylidene]-1,3-cyclohexanediol
IUPAC Name (1R,2R,3R,5Z)-5-[(2E)-2-[(1R,3aS,7aR)-1-[(1R)-5-hydroxy-1,5-dimethyl-hexyl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-2-(3-hydroxypropoxy)-4-methylene-cyclohexane-1,3-diol
SMILES C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)[C@@H](OCCCO)[C@H](O)C3=C)\CCC[C@]12C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C30H50O5/c1-20(9-6-15-29(3,4)34)24-13-14-25-22(10-7-16-30(24,25)5)11-12-23-19-26(32)28(27(33)21(23)2)35-18-8-17-31/h11-12,20,24-28,31-34H,2,6-10,13-19H2,1,3-5H3/b22-11+,23-12-/t20-,24-,25+,26-,27-,28-,30-/m1/s1
Formula Weight 490.3658
Chemical Name or Material Eldecalcitol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.