missing translation for 'onlineSavingsMsg'
Learn More

Eldecalcitol, MedChemExpress

Product Code. 16491999
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491999 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16491999 Supplier Medchem Express Supplier No. HYA0020
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Eldecalcitol (ED-71) is an orally active vitamin D3 analogue, inhibits bone resorption and increases bone mineral density. Eldecalcitol (ED-71) displays anti-tumor effect and inhibits cell proliferation, migration and induces apoptosis by suppressing GPx-1.

Eldecalcitol (ED-71) is an orally active vitamin D3 analogue, inhibits bone resorption and increases bone mineral density. Eldecalcitol (ED-71) displays anti-tumor effect and inhibits cell proliferation, migration and induces apoptosis by suppressing GPx-1.

Specifications

Chemical Name or Material Eldecalcitol
CAS 104121-92-8
Purity Grade Notes Research
Molecular Formula C30H50O5
Quantity 1 mg
Synonym ED-71 2-(3-hydroxypropoxy)-1,25-dihydroxyvitamin D3
Solubility Information DMSO : 3.33 mg/mL (6.79 mM; ultrasonic and warming and heat to 60°C)
SMILES O[C@H]1[C@@H](OCCCO)[C@H](O)C(/C(C1)=C\C=C2[C@]3([H])CC[C@H]([C@H](C)CCCC(C)(O)C)[C@@]3(C)CCC\2)=C
Molecular Weight (g/mol) 490.72
Formula Weight 490.72
Percent Purity 98.42%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.