missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELAVL3, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00001995-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MVTQILGAMESQVGGGPAGPALPNGPLLGTNGATDDSKTNLIVNYLPQNMTQDEFKSLFGSIGDIESCKLVRDKITGQSLGYGFVNYSDPNDADKAIDTLNGLKLQTKTIKVSYARPSSASIRDANLYVSGLPRTMSQKEMEQLFSQYGRIITSRILVDQVTGVSRGVGFIRFDKRIEAEEAIKGLNGQKPLGAAEPITVKFANNPSQKTGQALLTHLYQSSARRYAGPLHHQTQRFRLDNLLNMAYGVKSPLSLIARFSPIAIDGMSGLAGVGLSGGAAGAGWCIFVYNLSPEADESVLWQLFGPFGAVTNVKVIRDFTTNKCKGFGFVTMTNYDEAAMAIASLNGYRLGERVLQVSFKTSKQHKASpecifications
| ELAVL3 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| BC014144 | |
| ELAVL3 | |
| ELAVL3 (AAH11875, 1 a.a. ∼ 367 a.a) full-length human protein. | |
| RUO | |
| 1995 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| ELAV (embryonic lethal, abnormal vision, Drosophila)-like 3 (Hu antigen C) | |
| DKFZp547J036/HUC/HUCL/MGC20653/PLE21 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human, Rat | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction