missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Elastin Recombinant Rabbit Monoclonal Antibody (9F5R1)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558057

 View more versions of this product

Product Code. 30282497

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GGVPGAIPGG VPGGVFYPGA GLGALGGGAL GPGGKPLKPV PGGLAGAGLG AGLGAFPAVT FPGALVPGGV ADAAAAYKAA KAGAGLGGVP GVGGLGVSAG AVVPQPGAGV KPGKVPGVGL PGVYPGGVLP GARFPGVGVL PGVPTGAGVK PKAPGVGGAF AGIPGVGPFG GPQP.

This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal dominant cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Elastin
Recombinant Monoclonal
0.41 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P15502
ELN
Recombinant protein (or fragment).
20 μL
Primary
Human
Antibody
IgG
ELISA, Western Blot
9F5R1
Unconjugated
ELN
5830419M17Rik; AI385707; AI480567; AW229038; BB105748; E030024M20Rik; elastin; elastin (supravalvular aortic stenosis, Williams-Beuren syndrome); elastin a precursor; elastin b precursor; elastin c precursor; elastin microfibril interface located protein 1; elastin microfibril interface-located protein 1; Elastin microfibril interfacer 1; Eln; EMI; EMILIN; EMILIN1; EMILIN-1; FLJ38671; FLJ43523; gp115; RATTREL11; SVAS; TREL11; Trela; Trela26; Tropoelastin; WBS; WS
Rabbit
Protein A
RUO
2006
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.