missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elastin Mouse anti-Human, Clone: ELN/2069, Novus Biologicals™
Mouse Monoclonal Antibody
£396.00
Specifications
| Antigen | Elastin |
|---|---|
| Clone | ELN/2069 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
Description
Elastin Monoclonal specifically detects Elastin in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.Specifications
| Elastin | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Extracellular Matrix | |
| 10 mM PBS with 0.05% BSA | |
| 2006 | |
| IgG1 κ | |
| Protein A or G purified |
| ELN/2069 | |
| ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| elastin, FLJ38671, FLJ43523, SVAS, tropoelastin, WBS, WS | |
| Recombinant full-length human Elastin protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title