missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elastatinal, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.50mg
Specifications
Specifications
| Chemical Name or Material | Elastatinal |
| CAS | 51798-45-9 |
| Molecular Formula | C21 H36 N8 O7 |
| Quantity | 2.5 mg |
| Synonym | (2S)-2-[(4S)-2-Amino-3,4,5,6-tetrahydro-4-pyrimidinyl]-N-[[[(1S)-1-carboxy-3-methylbutyl]amino]carbonyl]glycyl-N1-[(1S)-1-methyl-2-oxoethyl]-L-glutamamide, Elastatinal, (2S)-2-[(4S)-2-Amino-1,4,5,6-tetrahydro-4-pyrimidinyl]-N-[[[(1S)-1-carboxy-3-methylbutyl]amino]carbonyl]glycyl-N1-[(1S)-1-methyl-2-oxoethyl]-L-glutamamide |
| InChI Formula | InChI=1S/C21H36N8O7/c1-10(2)8-14(19(34)35)28-21(36)29-16(12-6-7-24-20(23)27-12)18(33)26-13(4-5-15(22)31)17(32)25-11(3)9-30/h9-14,16H,4-8H2,1-3H3,(H2,22,31)(H,25,32)(H,26,33)(H,34,35)(H3,23,24,27)(H2,28,29,36)/t11-,12-,13-,14-,16-/m0/s1 |
| SMILES | CC(C)C[C@H](NC(=O)N[C@@H]([C@@H]1CCN=C(N)N1)C(=O)N[C@@H](CCC(=O)N)C(=O)N[C@@H](C)C=O)C(=O)O |
| IUPAC Name | (2S)-2-[[(1S)-2-[[(2S)-5-amino-1,5-dioxo-1-[[(2S)-1-oxopropan-2-yl]amino]pentan-2-yl]amino]-1-[(6S)-2-amino-1,4,5,6-tetrahydropyrimidin-6-yl]-2-oxoethyl]carbamoylamino]-4-methylpentanoic acid |
| Molecular Weight (g/mol) | 512.56 |
| Formula Weight | 512.271 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction