missing translation for 'onlineSavingsMsg'
Learn More

Elastatinal, Thermo Scientific Chemicals

Product Code. 15453769
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453769 5 mg 5mg
15463769 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453769 Supplier Thermo Scientific Alfa Aesar Supplier No. J61120.#0
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Irreversible elastase inhibitor

Irreversible elastase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51798-45-9
Molecular Formula C21H36N8O7
Molecular Weight (g/mol) 512.568
MDL Number MFCD00151180
InChI Key IJWCGVPEDDQUDE-WXZHEDMLSA-N
Synonym elastatinal, leu-co-cpd-gln-ala-h
PubChem CID 15942859
IUPAC Name (2S)-2-[[(1S)-2-[[(2S)-5-amino-1,5-dioxo-1-[[(2S)-1-oxopropan-2-yl]amino]pentan-2-yl]amino]-1-(2-amino-1,4,5,6-tetrahydropyrimidin-6-yl)-2-oxoethyl]carbamoylamino]-4-methylpentanoic acid
SMILES CC(C)CC(C(=O)O)NC(=O)NC(C1CCN=C(N1)N)C(=O)NC(CCC(=O)N)C(=O)NC(C)C=O

Specifications

Color White to Yellow
Quantity 5 mg
Beilstein 873543
Formula Weight 512.6
Physical Form Powder
Chemical Name or Material Elastatinal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.