missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Elastase Substrate V

Product Code. 15442929
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15442929 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15442929 Supplier Thermo Scientific Chemicals Supplier No. J60093.MB
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

627.70 g/mol

Fluorogenic substrate for quantification of human leukocyte and porcine pancreatic elastase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color White
Form Powder
For Use With (Application) Fluorogenic substrate for quantification of human leukocyte and porcine pancreatic elastase
Quantity 25 mg
Substrate Type Elastase Substrate
Molecular Weight (g/mol) 627.70
Storage Requirements -30° to -10°C
Substrate Quantity 25mg

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.