missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elastase Antibody (CELA3B/1757), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54491AF647
This item is not returnable.
View return policy
Description
Elastase Monoclonal antibody specifically detects Elastase in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF
Specifications
| Elastase | |
| Monoclonal | |
| Alexa Fluor 647 | |
| CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E | |
| Recombinant fragment (around aa 82-238) of human Elastase protein (exact sequence is proprietary) (Uniprot: P08861) | |
| 100 μg | |
| Cancer, Neuroscience | |
| 23436 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF | |
| CELA3B/1757 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction