missing translation for 'onlineSavingsMsg'
Learn More

Elaidic Acid, TRC

Product Code. 30368937
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30368937 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368937 Supplier Toronto Research Chemicals Supplier No. TRCE5002001G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Elaidic Acid
CAS 112-79-8
Molecular Formula C18 H34 O2
Quantity 1 g
Synonym 9-Octadecenoic acid, (E)-, Elaidic acid (8CI), (9E)-9-Octadecenoic acid, (9E)-Octadecenoic acid, (E)-9-Octadecenoic acid, (E)-Oleic acid, 9-trans-Octadecenoic acid, 9-trans-Oleic acid, Elaidinic acid, NSC 26988, trans-9-Octadecenoic acid, trans-Elaidic acid, trans-Oleic acid, trans-δ9-Octadecenoic acid, δ9-trans-Octadecenoic acid, Elaidic Acid, 9-Octadecenoic acid, (9E)-
InChI Formula InChI=1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9+
SMILES CCCCCCCC\C=C\CCCCCCCC(=O)O
IUPAC Name (E)-octadec-9-enoic acid
Molecular Weight (g/mol) 282.46
Formula Weight 282.2559
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.