missing translation for 'onlineSavingsMsg'
Learn More

Elaidic acid, 98%

Product Code. 11422597
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422597 25 g 25g
11402597 1 g 1g
11412597 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422597 Supplier Thermo Scientific Alfa Aesar Supplier No. A14832.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Store cold | CAS: 112-79-8 | C18H34O2 | 282.468 g/mol

In human platelets incubated with arachidonic acid, elaidic acid inhibits HHT and HETE formation while inducing prostaglandin and thromboxane synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In human platelets incubated with arachidonic acid, elaidic acid inhibits HHT and HETE formation while inducing prostaglandin and thromboxane synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-79-8
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.468
MDL Number MFCD00063954
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym elaidic acid, trans-oleic acid, trans-9-octadecenoic acid, 9-octadecenoic acid, e-oleic acid, 9-octadecenoic acid, e, e-octadec-9-enoic acid, trans-elaidic acid, 9-octadecenoic acid, 9e, elaidinsaure
PubChem CID 637517
ChEBI CHEBI:27997
IUPAC Name (E)-octadec-9-enoic acid
SMILES CCCCCCCCC=CCCCCCCCC(=O)O

Specifications

Melting Point 41°C to 45°C
Boiling Point 288°C (100 mmHg)
Flash Point >110°C (230°F)
Assay Percent Range 98%
Quantity 25 g
Beilstein 1726543
Merck Index 14,3533
Solubility Information Insoluble in water.
Formula Weight 282.47
Percent Purity 98%
Chemical Name or Material Elaidic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.