missing translation for 'onlineSavingsMsg'
Learn More

Elaidamide, TRC

Product Code. 30383831
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383831 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383831 Supplier Toronto Research Chemicals Supplier No. TRCE500000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Elaidamide
CAS 4303-70-2
Molecular Formula C18 H35 N O
Quantity 100 mg
Synonym 9-Octadecenamide, (9E)- (9CI, ACI), (9E)-9-Octadecenamide (ACI), 9-Octadecenamide, (E)- (ZCI), Elaidamide (6CI, 7CI, 8CI), (E)-9,10-Octadecenamide, Elaidic acid amide, Elaidic amide, Elaidoylamide, trans-9,10-Octadecenamide, trans-9-Octadecenamide
InChI Formula InChI=1S/C18H35NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H2,19,20)/b10-9+
SMILES CCCCCCCC\C=C\CCCCCCCC(=O)N
IUPAC Name (E)-octadec-9-enamide
Molecular Weight (g/mol) 281.48
Formula Weight 281.2719
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.