missing translation for 'onlineSavingsMsg'
Learn More

Elagolix Sodium, TRC

Product Code. 30381456
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381456 100 mg
30380351 500 mg
30387631 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381456 Supplier Toronto Research Chemicals Supplier No. TRCE501018100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 832720-36-2
Molecular Formula C32 H29 F5 N3 O5 . Na
Molecular Weight (g/mol) 653.57
Synonym Butanoic acid, 4-[[(1R)-2-[5-(2-fluoro-3-methoxyphenyl)-3-[[2-fluoro-6-(trifluoromethyl)phenyl]methyl]-3,6-dihydro-4-methyl-2,6-dioxo-1(2H)-pyrimidinyl]-1-phenylethyl]amino]-, sodium salt (1:1), Butanoic acid, 4-[[(1R)-2-[5-(2-fluoro-3-methoxyphenyl)-3-[[2-fluoro-6-(trifluoromethyl)phenyl]methyl]-3,6-dihydro-4-methyl-2,6-dioxo-1(2H)-pyrimidinyl]-1-phenylethyl]amino]-, monosodium salt (9CI), Elagolix sodium, NBI 56418NA, 4-[[(1R)-2-[5-(2-Fluoro-3-methoxyphenyl)-3-[[2-fluoro-6-(trifluoromethyl)phenyl]methyl]-3,6-dihydro-4-methyl-2,6-dioxo-1(2H)-pyrimidinyl]-1-phenylethyl]amino]butanoic acid sodium salt, (R)-4-((2-(5-(2-Fluoro-3-methoxyphenyl)-3-(2-fluoro-6-(trifluoromethyl)benzyl)-4-methyl-2,6-dioxo-2,3-dihydropyrimidin-1(6H)-yl)-1-phenylethyl)amino)butanoic acid sodium salt
IUPAC Name sodium;4-[[(1R)-2-[5-(2-fluoro-3-methoxyphenyl)-3-[[2-fluoro-6-(trifluoromethyl)phenyl]methyl]-4-methyl-2,6-dioxopyrimidin-1-yl]-1-phenylethyl]amino]butanoate
SMILES [Na+].COc1cccc(C2=C(C)N(Cc3c(F)cccc3C(F)(F)F)C(=O)N(C[C@H](NCCCC(=O)[O-])c4ccccc4)C2=O)c1F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C32H30F5N3O5.Na/c1-19-28(21-11-6-14-26(45-2)29(21)34)30(43)40(18-25(20-9-4-3-5-10-20)38-16-8-15-27(41)42)31(44)39(19)17-22-23(32(35,36)37)12-7-13-24(22)33;/h3-7,9-14,25,38H,8,15-18H2,1-2H3,(H,41,42);/q;+1/p-1/t25-;/m0./s1
Formula Weight 653.1925
Chemical Name or Material Elagolix Sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.