missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elagolix sodium, MedChemExpress
Description
Elagolix sodium is a human GnRH receptor (GnRHR) antagonist with an IC50 and Ki of 0.25 and 3.7 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | Elagolix sodium |
| CAS | 832720-36-2 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C32H29F5N3NaO5 |
| Quantity | 100 mg |
| Synonym | NBI-56418 sodium |
| Solubility Information | H2O : ≥ 50 mg/mL (76.50 mM) ∣DMSO : 50 mg/mL (76.50 mM; Need ultrasonic) |
| SMILES | O=C(O[Na])CCCN[C@H](C1=CC=CC=C1)CN(C(N(CC2=C(C(F)(F)F)C=CC=C2F)C(C)=C3C4=CC=CC(OC)=C4F)=O)C3=O |
| Molecular Weight (g/mol) | 653.57 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?