missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Elacytarabine

Product Code. 16446268
Encompass_Preferred
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 16446268

Brand: MedChemExpress HY14941

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Elacytarabine (CP 4055) is a lipid-conjugated derivative of the nucleoside analog cytarabine. Elacytarabine (CP 4055) is an antineoplastic drug with cytotoxicity in solid tumors.

Specifications

Molecular Weight (g/mol) 507.66 g/mol
Product Type Reference compound
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Purity 98.0%
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (246.23 mM∣Need ultrasonic)
For Use With (Application) Neuroscience-Neuromodulation
CAS 188181-42-2
Synonym CP 4055
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.