missing translation for 'onlineSavingsMsg'
Learn More

Elacridar, MedChemExpress

Product Code. 16481926
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481926 500 mg 500mg
16481326 200 mg 200mg
16458128 10 mg 10mg
16457886 100 mg 100mg
16414047 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16481926 Supplier Medchem Express Supplier No. HY50879
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Elacridar is an orally active P-glycoprotein (Pgp) and breast cancer resistance protein (BCRP) inhibitor. Elacridar can be used to examine the influence of efflux transporters on drug distribution to brain and the research of cancer.

Elacridar is an orally active P-glycoprotein (Pgp) and breast cancer resistance protein (BCRP) inhibitor. Elacridar can be used to examine the influence of efflux transporters on drug distribution to brain and the research of cancer.

Specifications

Chemical Name or Material Elacridar
CAS 143664-11-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C34H33N3O5
Quantity 500 mg
Synonym GF120918 GW0918 GG918 GW120918
Solubility Information DMSO : 5 mg/mL (8.87 mM; Need ultrasonic)
SMILES O=C1C2=CC=CC(C(NC(C=C3)=CC=C3CCN4CC(C=C(C(OC)=C5)OC)=C5CC4)=O)=C2NC6=C1C=CC=C6OC
Molecular Weight (g/mol) 563.64
Formula Weight 563.64
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.