missing translation for 'onlineSavingsMsg'
Learn More

Elacridar, MedChemExpress

Product Code. 16458128
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16458128

Brand: Medchem Express HY50879

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Elacridar is an orally active P-glycoprotein (Pgp) and breast cancer resistance protein (BCRP) inhibitor. Elacridar can be used to examine the influence of efflux transporters on drug distribution to brain and the research of cancer.

Elacridar is an orally active P-glycoprotein (Pgp) and breast cancer resistance protein (BCRP) inhibitor. Elacridar can be used to examine the influence of efflux transporters on drug distribution to brain and the research of cancer.

Specifications

Chemical Name or Material Elacridar
CAS 143664-11-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C34H33N3O5
Quantity 10 mg
Synonym GF120918 GW0918 GG918 GW120918
Solubility Information DMSO : 5 mg/mL (8.87 mM; Need ultrasonic)
SMILES O=C1C2=CC=CC(C(NC(C=C3)=CC=C3CCN4CC(C=C(C(OC)=C5)OC)=C5CC4)=O)=C2NC6=C1C=CC=C6OC
Molecular Weight (g/mol) 563.64
Formula Weight 563.64
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.