missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elacridar hydrochloride, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Elacridar hydrochloride (GF120918A) is an orally active P-glycoprotein (P-gp) and breast cancer resistance protein (BCRP) inhibitor. Elacridar hydrochloride can be used to examine the influence of efflux transporters on drug distribution to brain and it can be used for the research of cancer.
Specifications
Specifications
| Chemical Name or Material | Elacridar hydrochloride |
| CAS | 143851-98-3 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C34H34ClN3O5 |
| Quantity | 100 mg |
| Synonym | GF120918A |
| Solubility Information | DMSO : 62.5 mg/mL (104.15 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | O=C1C2=CC=CC(C(NC(C=C3)=CC=C3CCN4CC(C=C(C(OC)=C5)OC)=C5CC4)=O)=C2NC6=C1C=CC=C6OC.[H]Cl |
| Molecular Weight (g/mol) | 600.1 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction