missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elacridar hydrochloride, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Elacridar hydrochloride (GF120918A) is an orally active P-glycoprotein (P-gp) and breast cancer resistance protein (BCRP) inhibitor. Elacridar hydrochloride can be used to examine the influence of efflux transporters on drug distribution to brain and it can be used for the research of cancer.
Specifications
Specifications
| Chemical Name or Material | Elacridar hydrochloride |
| CAS | 143851-98-3 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C34H34ClN3O5 |
| Quantity | 10 mg |
| Synonym | GF120918A |
| Solubility Information | DMSO : 62.5 mg/mL (104.15 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | O=C1C2=CC=CC(C(NC(C=C3)=CC=C3CCN4CC(C=C(C(OC)=C5)OC)=C5CC4)=O)=C2NC6=C1C=CC=C6OC.[H]Cl |
| Molecular Weight (g/mol) | 600.1 |
| Vis mere |
Research purposes only
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion