missing translation for 'onlineSavingsMsg'
Learn More

Elacestrant dihydrochloride, MedChemExpress

Product Code. 16487928
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487928

Brand: Medchem Express HY19822A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Elacestrant dihydrochloride (RAD1901 dihydrochloride) is an orally available selective estrogen receptor degrader (SERD) with IC50s of 48 and 870 nM for ERα and ERβ, respectively.

Elacestrant dihydrochloride (RAD1901 dihydrochloride) is an orally available selective estrogen receptor degrader (SERD) with IC50s of 48 and 870 nM for ERα and ERβ, respectively.

Specifications

Chemical Name or Material Elacestrant dihydrochloride
CAS 1349723-93-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H40Cl2N2O2
Quantity 10 mg
Synonym RAD1901 dihydrochloride
Solubility Information DMSO : 100 mg/mL (188.13 mM; Need ultrasonic) ∣H2O : 50 mg/mL (94.06 mM; Need ultrasonic)
SMILES OC1=CC=C2C[C@H](C3=CC=C(OC)C=C3N(CC)CC4=CC=C(CCNCC)C=C4)CCC2=C1.[H]Cl.[H]Cl
Molecular Weight (g/mol) 531.56
Formula Weight 531.56
Percent Purity 95.73%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.