missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ELAC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30514
This item is not returnable.
View return policy
Description
ELAC2 Polyclonal antibody specifically detects ELAC2 in Human,Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| ELAC2 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| EC 3.1.26.11, elaC homolog 2 (E. coli), ElaC homolog protein 2, ELC2, FLJ10530, FLJ36693, FLJ42848, Heredity prostate cancer protein 2, HPC2elaC (E. coli) homolog 2, putative prostate cancer susceptibility protein HPC2/ELAC2, Ribonuclease Z 2, RNase Z 2, tRNA 3 endonuclease 2, tRNase Z (long form), tRNase Z 2, zinc phosphodiesterase ELAC protein 2 | |
| Between 776 and 826 | |
| 100 μg | |
| Cancer, Cell Biology, Cell Cycle and Replication | |
| 60528 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction