missing translation for 'onlineSavingsMsg'
Learn More

ELAC1 Antibody (1G2), Novus Biologicals™

Product Code. 18357979 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18357979 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18357979 Supplier Novus Biologicals Supplier No. H00055520M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ELAC1 Monoclonal antibody specifically detects ELAC1 in Human, Rat samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELAC1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1G2
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_061166
Gene Alias D29elaC (E. coli) homolog 1, Deleted in Ma29, EC 3.1.26.11, elaC homolog 1 (E. coli), ElaC homolog protein 1, FLJ59261, Ribonuclease Z 1, RNase Z 1, tRNA 3 endonuclease 1, tRNA 3' processing endoribonuclease, tRNA Z (short form), tRNase Z 1, tRNase ZS, zinc phosphodiesterase ELAC protein 1
Host Species Mouse
Immunogen ELAC1 (NP_061166, 282 a.a. ~ 362 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. QMDKAKEHGHSTPQMAATFAKLCRAKRLVLTHFSQRYKPVALAREGETDGIAELKKQAESVLDLQEVTLAEDFMVISIPIK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 55520
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.