Learn More
Invitrogen™ Total Antioxidant Capacity (T-AOC) Colorimetric Assay Kit
These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Brand: Invitrogen™ EEA022
Additional Details : Weight : 0.40000kg
Description
The Total Antioxidant Capacity (T-AOC) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of T-AOC in serum, plasma, whole blood, tissue, cells, and culture supernatant. T-AOC refers to the overall ability of a biological sample to counteract oxidative stress by neutralizing reactive oxygen species (ROS) and other harmful free radicals. It represents the cumulative effect of both enzymatic and non-enzymatic antioxidants present in the sample.
The assay quantifies the ability of the sample to reduce ferric ions (Fe3+) to ferrous ions (Fe2+) using a phenanthroline substance. The resulting complex exhibits a characteristic absorbance at 520 nm, which is directly proportional to the T-AOC of the sample.
Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results
Specifications
0.62–190.43 U/mL | |
Store at 2–8°C. |
|
Cell Culture Supernatant, Plasma, Serum, Tissue Homogenates, Whole Blood, Whole Cells | |
40 min. | |
4.8 | |
Dry Ice, 2–8°C | |
50 min. |
0.62 U/mL | |
Colorimetric | |
Microplate Reader | |
5.6 | |
20 μL | |
Total Antioxidant Capacity (T-AOC) |