Learn More
Invitrogen™ Low-density Lipoprotein Cholesterol (LDL-C) Colorimetric Assay Kit
Description
The Low-Density Lipoprotein Cholesterol (LDL-C) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of LDL-C in serum, plasma, tissue, cells, and cell culture supernatant samples. LDL-C is the main lipoprotein in fasting plasma, accounting for about two-thirds of plasma lipoproteins, and is the main vehicle for transporting cholesterol to extrahepatic tissues. The content of LDL-C is related to the incidence of cardiovascular disease and the degree of lesions and is considered to be the main pathogenic factor of atherosclerosis.
The assay is based on the reaction of LDL-C with enzyme reagents containing cholesterol esterase and cholesterol oxidase to produce hydrogen peroxide. Hydrogen peroxide is catalyzed by an oxidase in the presence of 4-aminoantipyrine (4-AA) and phenol (T-OOS) to form a red quinone compound with a maximum absorption peak at 546 nm. The LDL-C content in the sample can be calculated by measuring the OD value.
Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results
Specifications
Specifications
| Assay Range | 0.2–12 mmol/L |
| Assay Sensitivity | 0.2 mmol/L |
| Content And Storage | Store at 2–8°C. |
| Detection Method | Colorimetric |
| Sample Type | Cell Culture Supernatant, Plasma, Serum, Tissue Homogenates, Whole Cells |
| For Use With (Equipment) | Microplate Reader |
| Incubation Time | 10 min. |
| Interassay CV | 10 |
| Intraassay CV | 5.5 |
| Quantity | 96 tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.