missing translation for 'onlineSavingsMsg'
Learn More

ELA3A Antibody, Novus Biologicals™

Product Code. 18369029 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369029 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369029 Supplier Novus Biologicals Supplier No. H00010136B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

ELA3A Polyclonal antibody specifically detects ELA3A in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELA3A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_005738.3
Gene Alias chymotrypsin-like elastase family member 3A, chymotrypsin-like elastase family, member 3A, EC 3.4.21, EC 3.4.21.70, ELA3A, ELA3elastase 1, elastase 3A, pancreatic, elastase 3A, pancreatic (protease E), Elastase IIIA, elastase-3A, Protease E
Host Species Mouse
Immunogen ELA3A (NP_005738.3, 1 a.a. - 270 a.a.) full-length human protein. MMLRLLSSLLLVAVASGYGPPSSHSSSRVVHGEDAVPYSWPWQVSLQYEKSGSFYHTCGGSLIAPDWVVTAGHCISRDLTYQVVLGEYNLAVKEGPEQVIPINSEELFVHPLWNRSCVACGNDIALIKLSRSAQLGDAVQLASLPPAGDILPNKTPCYITGWGRLYTNGPLPDKLQQARLPVVDYKHCSRWNWWGSTVKKTMVCAGGYIRSGCNGDSGGPLNCPTEDGGWQVHGVTSFVSGFGCNFIWKPTVFTRVSAFIDWIEETIASH
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 10136
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.