missing translation for 'onlineSavingsMsg'
Learn More

ELA2A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18384646 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384646 25 μg 25µL
18399966 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18384646 Supplier Novus Biologicals Supplier No. NBP31064525UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ELA2A Polyclonal specifically detects ELA2A in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELA2A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias chymotrypsin-like elastase family member 2A, chymotrypsin-like elastase family, member 2A, EC 3.4.21, EC 3.4.21.71, ELA2Apancreatic elastase 2, elastase 2A, Elastase-2A, pancreatic elastase IIA, PE-1
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of human ELA2A (NP_254275.1). Peptide sequence TGWGRLQTNGAVPDVLQQGRLLVVDYATCSSSAWWGSSVKTSMICAGGDG
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63036
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.