missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELA1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55342-1-AP-20UL
This item is not returnable.
View return policy
Description
The ELA1 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human ELA1. This antibody recognizes human, mouse antigen. The ELA1 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| ELA1 | |
| Polyclonal | |
| Unconjugated | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 28 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 600 μg/mL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1990 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction