missing translation for 'onlineSavingsMsg'
Learn More

Manutan Ekwo bubble film. Lxw 150m x 1500mm. Bubb dia 10mm

Brand:  Manutan A027094

Encompass_Preferred

Ekwo bubble film. Lxw 150m x 1500mm. Bubb dia 10mm

Product Code. 12868339

  • £162.75 / Pack of 1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.