Learn More
Invitrogen™ EKLF Recombinant Superclonal™ Antibody (6HCLC)
Rabbit Recombinant Superclonal Antibody
Brand: Invitrogen™ 710917
Description
This antibody is predicted to react with Monkey . Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
KLF1 is a hematopoietic-specific transcription factor that induces high-level expression of adult beta-globin and other erythroid genes. The zinc-finger protein binds to the DNA sequence CCACACCCT found in the beta hemoglobin promoter. Heterozygous loss-of-function mutations in this gene result in the dominant Inblood phenotype.
Specifications
| EKLF | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q13351 | |
| Klf1 | |
| Multiple peptides corresponding to Human KLF1 (aa 40-57; 253-269). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 6HCLC | |
| Unconjugated | |
| Klf1 | |
| CDAN4; Eklf; Erythroid krueppel-like transcription factor; erythroid Kruppel-like factor; erythroid transcription factor; erythroid-specific transcription factor EKLF; HBFQTL6; INLU; KLF1; Krueppel-like factor 1; Kruppel like factor 1; Kruppel-like factor 1 (erythroid); monoclonal antibody A3D8; Nan; neonatal anemia; truncated EKLF; truncated erythroid Kruppel-like factor 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10661 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.