missing translation for 'onlineSavingsMsg'
Learn More

EKASTU Safety EKASTU Mask Disinfectant

Product Code. 10549432
Change view
Click to view available options
Height (Metric):
4.5 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric)
10549432 4.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10549432 Supplier EKASTU Safety Supplier No. 266942
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EKASTU Mask Disinfectant is an effective long-term disinfectant for masks

  • Formaldehyde-free
  • Pleasant smell
  • Will not stain surface
  • Non-stick valves
  • 250mL (concentrate)

Specifications

Height (Metric) 4.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.