Learn More
EISCO Spectrum Tube Power Supply
Brand:  EISCO PH1197A
Product Code. 13102990
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Black panel behind the tube eliminates distracting ambient light and protects tube from breakage
  • Specially designed box holds tube firmly, preventing student from touching electrodes
  • Suitable for 10.25 in. (26cm) long spectrum tubes
Order Info
Operates on 120VAC 60Hz ±10%.