Learn More
EISCO Magnetic Field Demonstrator
Designed to show configuration of a magnetic field in one or two dimensions.
Brand:  EISCO PH0803
Product Code. 13166490
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes: Instructions for use
Description
Description
  • Consists of a set of two transparent plastic plates, 6.0 × 3 × 0.24 in. (153 × 76 × 6mm)
  • Each plate has a matrix of 98 small circular chambers, 0.5 × 0.3 in. (14 × 7mm)
  • The transparent nature of the plates makes them suitable for use on an overhead projector